Product Details

SNP ID
rs138609657
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124440399 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTTTGGACCTGCCCTCCGTGGA[A/G]TCAATGTGGAAGATGCTGGAGAGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR8B8 PubMed Links

Gene Details

Gene
OR8B8
Gene Name
olfactory receptor family 8 subfamily B member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012378.1 687 Silent Mutation GAC,GAT D229D NP_036510.1

View Full Product Details