Product Details

SNP ID
rs138649062
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:396655 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCTGCAGGCTGGCTTCAGCTCT[C/T]GCTCTCAGGGCCTGAGTGGGGACAA
Phenotype
MIM: 605561 MIM: 605478
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PKP3 PubMed Links

Gene Details

Gene
PKP3
Gene Name
plakophilin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303029.1 390 Missense Mutation CGC,TGC R109C NP_001289958.1
NM_007183.3 390 Missense Mutation CGC,TGC R94C NP_009114.1
Gene
SIGIRR
Gene Name
single Ig and TIR domain containing
There are no transcripts associated with this gene.

View Full Product Details