Product Details

SNP ID
rs138744799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65031366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCAAGTACCCCAAACCATAGGA[C/T]ATTATTTTAAAATTATTATGAACAT
Phenotype
MIM: 601175 MIM: 605964
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL2 PubMed Links

Gene Details

Gene
ARL2
Gene Name
ADP ribosylation factor like GTPase 2
There are no transcripts associated with this gene.

Gene
ARL2-SNX15
Gene Name
ARL2-SNX15 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
SAC3D1
Gene Name
SAC3 domain containing 1
There are no transcripts associated with this gene.

Gene
SNX15
Gene Name
sorting nexin 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013306.4 Intron NP_037438.2
NM_147777.3 Intron NP_680086.2

View Full Product Details