Product Details

SNP ID
rs139340528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6795041 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCACAGCACCAGTAATGGCCACA[C/T]AGGAGGCCCCAGTGACAGAGAGTGG
Phenotype
MIM: 608495
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2AG1 PubMed Links

Gene Details

Gene
OR2AG1
Gene Name
olfactory receptor family 2 subfamily AG member 1 (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
OR6A2
Gene Name
olfactory receptor family 6 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003696.2 868 Missense Mutation TAT,TGT Y223C NP_003687.2

View Full Product Details