Product Details

SNP ID
rs139576982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:821676 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTATCTAAAGAGGCCCGGAAGC[A/G]GTTCCTGGGCCCCCTGCACCCCTCC
Phenotype
MIM: 614177 MIM: 609059 MIM: 180530
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRACR2B PubMed Links

Gene Details

Gene
CRACR2B
Gene Name
calcium release activated channel regulator 2B
There are no transcripts associated with this gene.

Gene
PNPLA2
Gene Name
patatin like phospholipase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020376.3 385 Missense Mutation CAG,CGG Q79R NP_065109.1
XM_006718265.3 385 Missense Mutation CAG,CGG Q79R XP_006718328.1
XM_006718266.3 385 Missense Mutation CAG,CGG Q79R XP_006718329.1
XM_017018028.1 385 Missense Mutation CAG,CGG Q79R XP_016873517.1
Gene
RPLP2
Gene Name
ribosomal protein lateral stalk subunit P2
There are no transcripts associated with this gene.

Gene
SNORA52
Gene Name
small nucleolar RNA, H/ACA box 52
There are no transcripts associated with this gene.

View Full Product Details