Product Details
- SNP ID
-
rs139700832
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:113323314 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAATTTAATTCAGGAGATGAATTCT[C/G]ATGACCCAGTTGTGCAACAGAAAGC
- Phenotype
-
MIM: 610732
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TTC12
PubMed Links
Gene Details
- Gene
- TTC12
- Gene Name
- tetratricopeptide repeat domain 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001318533.1 |
190 |
Missense Mutation |
CAT,GAT |
H29D |
NP_001305462.1 |
NM_017868.3 |
190 |
Missense Mutation |
CAT,GAT |
H29D |
NP_060338.3 |
XM_005271604.3 |
190 |
Missense Mutation |
CAT,GAT |
H29D |
XP_005271661.1 |
XM_011542890.2 |
190 |
UTR 5 |
|
|
XP_011541192.1 |
XM_017017943.1 |
190 |
Missense Mutation |
CAT,GAT |
H29D |
XP_016873432.1 |
XM_017017944.1 |
190 |
Missense Mutation |
CAT,GAT |
H29D |
XP_016873433.1 |
XM_017017945.1 |
190 |
Missense Mutation |
CAT,GAT |
H29D |
XP_016873434.1 |
XM_017017946.1 |
190 |
Missense Mutation |
CAT,GAT |
H29D |
XP_016873435.1 |
XM_017017947.1 |
190 |
Missense Mutation |
CAT,GAT |
H29D |
XP_016873436.1 |
XM_017017948.1 |
190 |
Missense Mutation |
CAT,GAT |
H29D |
XP_016873437.1 |
XM_017017949.1 |
190 |
Missense Mutation |
CAT,GAT |
H29D |
XP_016873438.1 |
XM_017017950.1 |
190 |
Missense Mutation |
CAT,GAT |
H29D |
XP_016873439.1 |
View Full Product Details