Product Details

SNP ID
rs140056826
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55839291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCAACACAATCAATCATTTCTT[C/T]TGTGAGTTATCCTCCCTGATATCAC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR5D16 PubMed Links

Gene Details

Gene
OR5D16
Gene Name
olfactory receptor family 5 subfamily D member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005496.1 540 Silent Mutation TTC,TTT F180F NP_001005496.1

View Full Product Details