Product Details

SNP ID
rs140341027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118551843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCTGCATCAGCGTGTCAATGTC[A/G]GGCATGGGCCTAAAAGTATAAAGGT
Phenotype
MIM: 613934
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFT46 PubMed Links

Gene Details

Gene
IFT46
Gene Name
intraflagellar transport 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168618.1 1034 Silent Mutation CCC,CCT P205P NP_001162089.1
NM_020153.3 1034 Silent Mutation CCC,CCT P256P NP_064538.3
XM_011542905.2 1034 Silent Mutation CCC,CCT P256P XP_011541207.1
XM_011542906.2 1034 Silent Mutation CCC,CCT P205P XP_011541208.1
XM_017018017.1 1034 Silent Mutation CCC,CCT P256P XP_016873506.1
XM_017018018.1 1034 Silent Mutation CCC,CCT P205P XP_016873507.1
Gene
TMEM25
Gene Name
transmembrane protein 25
There are no transcripts associated with this gene.

View Full Product Details