Product Details

SNP ID
rs140467802
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124440374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATGTGGGAGCTGCAGGTGCTGA[A/T]GGCTTTGGACCTGCCCTCCGTGGAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR8B8 PubMed Links

Gene Details

Gene
OR8B8
Gene Name
olfactory receptor family 8 subfamily B member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012378.1 712 Missense Mutation ATC,TTC I238F NP_036510.1

View Full Product Details