Product Details

SNP ID
rs140567674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:69773449 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGCGTTGTAGTTGTTGGGAAGGA[A/G]AATCTCCTTGAACGTGCACTCATCG
Phenotype
MIM: 164980
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGF4 PubMed Links

Gene Details

Gene
FGF4
Gene Name
fibroblast growth factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002007.2 800 Missense Mutation CTC,TTC L161F NP_001998.1
XM_005273847.1 800 UTR 3 XP_005273904.1

View Full Product Details