Product Details

SNP ID
rs140587054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58228556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCGTGCTGCCGAGGGTGACAAAG[A/C]AGAACATTTGGGCCCCACATCCAGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR10Q1 PubMed Links

Gene Details

Gene
OR10Q1
Gene Name
olfactory receptor family 10 subfamily Q member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004471.2 363 Missense Mutation TGC,TTC C107F NP_001004471.1

View Full Product Details