Product Details

SNP ID
rs141003895
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55554592 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGCAACTGTTGGGGGCAACATG[C/T]TAATTGTAGTAACCATTCTCAGCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4C15 PubMed Links

Gene Details

Gene
OR4C15
Gene Name
olfactory receptor family 4 subfamily C member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001920.1 286 Silent Mutation CTA,TTA L96L NP_001001920.1

View Full Product Details