Product Details

SNP ID
rs141078675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57537782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTTTCTAATTCTCTCCACCACA[A/G]TGTCTACTCTCTTTAGATGCCTGGG
Phenotype
MIM: 613664 MIM: 602251
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMTNL1 PubMed Links

Gene Details

Gene
SMTNL1
Gene Name
smoothelin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105565.2 Intron NP_001099035.2
XM_005273819.3 Intron XP_005273876.1
Gene
TIMM10
Gene Name
translocase of inner mitochondrial membrane 10 homolog (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012456.2 Intron NP_036588.1

View Full Product Details