Product Details

SNP ID
rs141177910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67435917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACTGTCTCTGGCTTCCTCAGCC[C/T]GCACGGGGACCTGCTCTGGGCTGGA
Phenotype
MIM: 609849 MIM: 601577 MIM: 608939
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CORO1B PubMed Links

Gene Details

Gene
CORO1B
Gene Name
coronin 1B
There are no transcripts associated with this gene.

Gene
PTPRCAP
Gene Name
protein tyrosine phosphatase, receptor type C associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005608.2 500 Missense Mutation CAG,CGG Q146R NP_005599.1
Gene
RPS6KB2
Gene Name
ribosomal protein S6 kinase B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003952.2 500 Intron NP_003943.2
XM_005274164.1 500 Intron XP_005274221.1
XM_005274165.4 500 Intron XP_005274222.1
XM_006718655.3 500 Intron XP_006718718.1
XM_006718656.3 500 Intron XP_006718719.1
XM_006718657.1 500 Intron XP_006718720.1
XM_017018108.1 500 Intron XP_016873597.1

View Full Product Details