Product Details

SNP ID
rs141189792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56232208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGTTCCTCAAACTGTAGATGAC[A/G]GGATTCAGCAAGGGAATCACAATGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5T2 PubMed Links

Gene Details

Gene
OR5T2
Gene Name
olfactory receptor family 5 subfamily T member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004746.1 978 Silent Mutation CCC,CCT P326P NP_001004746.1

View Full Product Details