Product Details

SNP ID
rs141345438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66564626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCTGCAGAGGGGCCGGAGAGGG[C/T]GGGAGATCCCGTGGCGGTAAAACTG
Phenotype
MIM: 102574 MIM: 603539
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTN3 PubMed Links

Gene Details

Gene
ACTN3
Gene Name
actinin alpha 3 (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
CTSF
Gene Name
cathepsin F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003793.3 1093 Missense Mutation CAC,CGC H418R NP_003784.2
XM_011545328.2 1093 Missense Mutation CAC,CGC H358R XP_011543630.1

View Full Product Details