Product Details

SNP ID
rs141519982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68157637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGTTGAGGAATAATTGACTGGA[A/G]TTTTTTATTTCTTTAAAGTAGTTAT
Phenotype
MIM: 610881
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KMT5B PubMed Links

Gene Details

Gene
KMT5B
Gene Name
lysine methyltransferase 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300907.1 3061 UTR 3 NP_001287836.1
NM_001300908.1 3061 UTR 3 NP_001287837.1
NM_001300909.1 3061 Intron NP_001287838.1
NM_016028.4 3061 Intron NP_057112.3
NM_017635.4 3061 UTR 3 NP_060105.3
XM_005274035.3 3061 UTR 3 XP_005274092.2
XM_005274036.3 3061 UTR 3 XP_005274093.2
XM_006718581.1 3061 UTR 3 XP_006718644.1
XM_011545091.1 3061 UTR 3 XP_011543393.1
XM_011545092.2 3061 UTR 3 XP_011543394.1
XM_011545093.2 3061 UTR 3 XP_011543395.1
XM_011545094.2 3061 Intron XP_011543396.1
XM_017017876.1 3061 UTR 3 XP_016873365.1
XM_017017877.1 3061 UTR 3 XP_016873366.1
XM_017017878.1 3061 UTR 3 XP_016873367.1
XM_017017879.1 3061 UTR 3 XP_016873368.1
XM_017017880.1 3061 Intron XP_016873369.1

View Full Product Details