Product Details

SNP ID
rs141720771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:26668573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTAGCTGAATCTATTGACTGTGC[A/G]CTTCATAGTTTCCTTTTGCTTCACC
Phenotype
MIM: 610110 MIM: 612455
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANO3 PubMed Links

Gene Details

Gene
ANO3
Gene Name
anoctamin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001313726.1 4625 Intron NP_001300655.1
NM_001313727.1 4625 Intron NP_001300656.1
NM_031418.3 4625 Intron NP_113606.2
Gene
SLC5A12
Gene Name
solute carrier family 5 member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178498.3 4625 UTR 3 NP_848593.2
XM_006718155.3 4625 UTR 3 XP_006718218.1
XM_006718156.3 4625 Intron XP_006718219.1
XM_011519920.2 4625 UTR 3 XP_011518222.1
XM_011519921.2 4625 Intron XP_011518223.1
XM_017017244.1 4625 UTR 3 XP_016872733.1

View Full Product Details