Product Details

SNP ID
rs141745866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59364718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCCAAGATGCTCTCCAACCTCT[G/T]ACAGGAACAGCAAACTATCACTTTT
Phenotype
MIM: 615702
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR5AN1 PubMed Links

Gene Details

Gene
OR5AN1
Gene Name
olfactory receptor family 5 subfamily AN member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004729.1 260 Nonsense Mutation TGA,TTA *87L NP_001004729.1

View Full Product Details