Product Details

SNP ID
rs141888730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60937241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCAGCCCCTGTCCCTGCTGAAA[C/T]GGCTGTGGGAGGCTGGGCCCTGGGA
Phenotype
MIM: 610408
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC15A3 PubMed Links

Gene Details

Gene
SLC15A3
Gene Name
solute carrier family 15 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016582.2 1958 Missense Mutation CAT,CGT H575R NP_057666.1
XM_011545095.2 1958 UTR 3 XP_011543397.1
Gene
TMEM132A
Gene Name
transmembrane protein 132A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017870.3 1958 Intron NP_060340.2
NM_178031.2 1958 Intron NP_821174.1
XM_017017951.1 1958 Intron XP_016873440.1
XM_017017952.1 1958 Intron XP_016873441.1

View Full Product Details