Product Details

SNP ID
rs141987293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60869565 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGTTTCCACCTGAGGGTGTTCA[C/T]GGAGGCTGTGCTGCCCAATGGTCGT
Phenotype
MIM: 195000
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZP1 PubMed Links

Gene Details

Gene
ZP1
Gene Name
zona pellucida glycoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207341.3 367 Missense Mutation ACG,ATG T116M NP_997224.2
XM_011544852.2 367 Intron XP_011543154.1
XM_011544853.2 367 Intron XP_011543155.1

View Full Product Details