Product Details

SNP ID
rs142055221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58228700 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAAGAAATACATCGGGGTGCGG[A/T]GGGTGCTGTGTGTGCACACCACCCA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR10Q1 PubMed Links

Gene Details

Gene
OR10Q1
Gene Name
olfactory receptor family 10 subfamily Q member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004471.2 219 Missense Mutation CAC,CTC H59L NP_001004471.1

View Full Product Details