Product Details

SNP ID
rs142257440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:10305718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCGGGATGAAGCTGGTTTCCGT[C/T]GCCCTGATGTACCTGGGTTCGCTCG
Phenotype
MIM: 103275
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADM PubMed Links

Gene Details

Gene
ADM
Gene Name
adrenomedullin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001124.2 289 Silent Mutation GTC,GTT V6V NP_001115.1
Gene
CAND1.11
Gene Name
uncharacterized LOC100130460
There are no transcripts associated with this gene.

Gene
LOC105376544
Gene Name
uncharacterized LOC105376544
There are no transcripts associated with this gene.

View Full Product Details