Product Details
- SNP ID
-
rs142344804
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:9021092 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGACACTTTGGAACGTAGCAATC[A/G]GATGAACGATCTTGGAAACATCTCT
- Phenotype
-
MIM: 611747
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SCUBE2
PubMed Links
Gene Details
- Gene
- SCUBE2
- Gene Name
- signal peptide, CUB domain and EGF like domain containing 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001170690.1 |
3111 |
Nonsense Mutation |
CGA,TGA |
R793* |
NP_001164161.1 |
NM_020974.2 |
3111 |
Nonsense Mutation |
CGA,TGA |
R957* |
NP_066025.2 |
XM_005253032.4 |
3111 |
Nonsense Mutation |
CGA,TGA |
R1014* |
XP_005253089.2 |
XM_005253033.4 |
3111 |
Nonsense Mutation |
CGA,TGA |
R985* |
XP_005253090.2 |
XM_005253034.4 |
3111 |
Nonsense Mutation |
CGA,TGA |
R973* |
XP_005253091.2 |
XM_005253035.4 |
3111 |
Intron |
|
|
XP_005253092.2 |
XM_005253036.4 |
3111 |
Nonsense Mutation |
CGA,TGA |
R905* |
XP_005253093.2 |
XM_005253037.4 |
3111 |
Nonsense Mutation |
CGA,TGA |
R859* |
XP_005253094.2 |
XM_011520246.2 |
3111 |
Nonsense Mutation |
CGA,TGA |
R948* |
XP_011518548.1 |
XM_011520247.2 |
3111 |
Nonsense Mutation |
CGA,TGA |
R946* |
XP_011518549.1 |
XM_011520248.2 |
3111 |
Intron |
|
|
XP_011518550.1 |
XM_017018080.1 |
3111 |
Nonsense Mutation |
CGA,TGA |
R944* |
XP_016873569.1 |
XM_017018081.1 |
3111 |
Nonsense Mutation |
CGA,TGA |
R878* |
XP_016873570.1 |
XM_017018082.1 |
3111 |
Intron |
|
|
XP_016873571.1 |
View Full Product Details