Product Details

SNP ID
rs142457418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62707671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGGTATGCTCCAGGATCTGAGC[A/C]GCTCCTTCTAGCATCCTTCATCCTT
Phenotype
MIM: 606158 MIM: 608941
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BSCL2 PubMed Links

Gene Details

Gene
BSCL2
Gene Name
BSCL2, seipin lipid droplet biogenesis associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122955.3 78 UTR 5 NP_001116427.1
NM_001130702.2 78 Intron NP_001124174.2
NM_032667.6 78 Intron NP_116056.3
Gene
GNG3
Gene Name
G protein subunit gamma 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012202.4 78 UTR 5 NP_036334.1
XM_006718500.2 78 Intron XP_006718563.1
Gene
HNRNPUL2
Gene Name
heterogeneous nuclear ribonucleoprotein U like 2
There are no transcripts associated with this gene.

Gene
HNRNPUL2-BSCL2
Gene Name
HNRNPUL2-BSCL2 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details