Product Details

SNP ID
rs142523550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66564134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCTGTTCTTGATGGCCCAAAAG[A/G]GAACGTCAGAGCCTGGGGTGCAGTG
Phenotype
MIM: 102574 MIM: 603539
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTN3 PubMed Links

Gene Details

Gene
ACTN3
Gene Name
actinin alpha 3 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104.3 1174 Intron NP_001095.2
NM_001258371.2 1174 Intron NP_001245300.2
Gene
CTSF
Gene Name
cathepsin F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003793.3 1174 Missense Mutation CCC,CTC P445L NP_003784.2
XM_011545328.2 1174 Missense Mutation CCC,CTC P385L XP_011543630.1

View Full Product Details