Product Details
- SNP ID
-
rs142595009
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:4489028 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTGCTTATACTCTGGCCCTGCTAG[C/G]CAACTGTACCCTTCTCTTCATTATC
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR52K1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs96489] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR52K1
- Gene Name
- olfactory receptor family 52 subfamily K member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001005171.2 |
150 |
Missense Mutation |
GCC,GGC |
A43G |
NP_001005171.2 |
View Full Product Details