Product Details

SNP ID
rs142853719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:69295679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGGCTCCAAAGCCGGCAGATCC[C/T]GGGGCCGCCTCTGTCTCTCCCAGGC
Phenotype
MIM: 605625
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYEOV PubMed Links

Gene Details

Gene
MYEOV
Gene Name
myeloma overexpressed
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293291.1 527 Missense Mutation CGG,TGG R77W NP_001280220.1
NM_001293294.1 527 Missense Mutation CGG,TGG R19W NP_001280223.1
NM_001293296.1 527 Missense Mutation CGG,TGG R19W NP_001280225.1
NM_001300923.1 527 Missense Mutation CGG,TGG R19W NP_001287852.1
NM_001300924.1 527 Missense Mutation CGG,TGG R19W NP_001287853.1
NM_138768.3 527 Missense Mutation CGG,TGG R77W NP_620123.2

View Full Product Details