Product Details

SNP ID
rs142861804
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:792341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACGGCCACAGCGGCGGCACTCCC[A/G]GCCACACAGCCGGCCAGGAAGGACA
Phenotype
MIM: 608213 MIM: 605247 MIM: 609302
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEND1 PubMed Links

Gene Details

Gene
CEND1
Gene Name
cell cycle exit and neuronal differentiation 1
There are no transcripts associated with this gene.

Gene
LOC171391
Gene Name
uncharacterized LOC171391
There are no transcripts associated with this gene.

Gene
PANO1
Gene Name
proapoptotic nucleolar protein 1
There are no transcripts associated with this gene.

Gene
PIDD1
Gene Name
p53-induced death domain protein 1
There are no transcripts associated with this gene.

Gene
SLC25A22
Gene Name
solute carrier family 25 member 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191060.1 1031 Silent Mutation GCC,GCT A235A NP_001177989.1
NM_001191061.1 1031 Silent Mutation GCC,GCT A235A NP_001177990.1
NM_024698.5 1031 Silent Mutation GCC,GCT A235A NP_078974.1
XM_011520369.1 1031 Silent Mutation GCC,GCT A235A XP_011518671.1
XM_011520370.1 1031 Silent Mutation GCC,GCT A235A XP_011518672.1
XM_011520371.1 1031 Silent Mutation GCC,GCT A235A XP_011518673.1

View Full Product Details