Product Details

SNP ID
rs142871932
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:3639683 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTAAAATGGCTACAGGTCTGATTA[C/T]AGCTCCAGAGAGTCACCAAGCTCTG
Phenotype
MIM: 601625 MIM: 610625
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ART1 PubMed Links
Additional Information
For this assay, SNP(s) [rs2271589] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ART1
Gene Name
ADP-ribosyltransferase 1
There are no transcripts associated with this gene.

Gene
ART5
Gene Name
ADP-ribosyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079536.1 387 Missense Mutation TAT,TGT Y249C NP_001073004.1
NM_001297668.1 387 Intron NP_001284597.1
NM_053017.4 387 Missense Mutation TAT,TGT Y249C NP_443750.2
XM_005252779.3 387 Missense Mutation TAT,TGT Y249C XP_005252836.1
XM_006718136.3 387 Missense Mutation TAT,TGT Y249C XP_006718199.1
XM_011519879.2 387 Missense Mutation TAT,TGT Y249C XP_011518181.1
XM_017017171.1 387 Missense Mutation TAT,TGT Y57C XP_016872660.1
Gene
TRPC2
Gene Name
transient receptor potential cation channel subfamily C member 2, pseudogene
There are no transcripts associated with this gene.

View Full Product Details