Product Details

SNP ID
rs143065051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5544712 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCGAGAGACATTGTGTCCAAAG[C/T]GATGGGCGAGGATGGAGAAAAAGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR52H1 PubMed Links

Gene Details

Gene
OR52H1
Gene Name
olfactory receptor family 52 subfamily H member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005289.1 812 Missense Mutation CAC,CGC H271R NP_001005289.1

View Full Product Details