Product Details

SNP ID
rs143088995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47605253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCCCATATTTTTCACTGTTTCA[A/C]CCCTTTGTAACTAAAATAACAAGAG
Phenotype
MIM: 614911 MIM: 613221
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C1QTNF4 PubMed Links

Gene Details

Gene
C1QTNF4
Gene Name
C1q and tumor necrosis factor related protein 4
There are no transcripts associated with this gene.

Gene
MTCH2
Gene Name
mitochondrial carrier 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317231.1 1959 Intron NP_001304160.1
NM_001317232.1 1959 Intron NP_001304161.1
NM_001317233.1 1959 Intron NP_001304162.1
NM_014342.3 1959 Intron NP_055157.1
XM_006718172.2 1959 Intron XP_006718235.1
XM_011519959.2 1959 UTR 3 XP_011518261.1
XM_011519960.2 1959 UTR 3 XP_011518262.1
XM_011519961.2 1959 UTR 3 XP_011518263.1
XM_017017462.1 1959 Intron XP_016872951.1
XM_017017463.1 1959 Intron XP_016872952.1

View Full Product Details