Product Details

SNP ID
rs143102499
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:20364833 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTCAACCTGCGCACTTCACACT[A/G]ATATTGTCAGGGATTCTGAAACTGC
Phenotype
MIM: 605628
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HTATIP2 PubMed Links

Gene Details

Gene
HTATIP2
Gene Name
HIV-1 Tat interactive protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098520.1 Intron NP_001091990.1
NM_001098521.1 Intron NP_001091991.1
NM_001098522.1 Intron NP_001091992.1
NM_001098523.1 Intron NP_001091993.1
NM_006410.4 Intron NP_006401.3

View Full Product Details