Product Details

SNP ID
rs143479406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:69643953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGAGGAGAACAAACAGATCATCC[A/G]CAAACACGCGCAGACCTTCGTTGCC
Phenotype
MIM: 168461
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCND1 PubMed Links

Gene Details

Gene
CCND1
Gene Name
cyclin D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053056.2 745 Missense Mutation CAC,CGC H179R NP_444284.1

View Full Product Details