Product Details

SNP ID
rs144165811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67438478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCAGGGCCCGCAGCTCCTGCAT[C/T]ACCTCCTCCAGCTTCCCAGCCTCCT
Phenotype
MIM: 609849 MIM: 601577 MIM: 608939
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CORO1B PubMed Links

Gene Details

Gene
CORO1B
Gene Name
coronin 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018070.2 1479 Silent Mutation GTA,GTG V456V NP_001018080.1
NM_020441.2 1479 Silent Mutation GTA,GTG V456V NP_065174.1
Gene
PTPRCAP
Gene Name
protein tyrosine phosphatase, receptor type C associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005608.2 1479 Intron NP_005599.1
Gene
RPS6KB2
Gene Name
ribosomal protein S6 kinase B2
There are no transcripts associated with this gene.

View Full Product Details