Product Details

SNP ID
rs144434585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:128937677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCGGTTTTCACTGCAGGGACTTA[C/T]CCAGCCAGGTGTGTGTGTCTGAGCC
Phenotype
MIM: 605426
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TP53AIP1 PubMed Links

Gene Details

Gene
TP53AIP1
Gene Name
tumor protein p53 regulated apoptosis inducing protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195194.1 824 Intron NP_001182123.1
NM_001195195.1 824 Intron NP_001182124.1
NM_001251964.1 824 Missense Mutation ATA,GTA I48V NP_001238893.1
NM_022112.2 824 Intron NP_071395.2
XM_017018113.1 824 Intron XP_016873602.1
XM_017018114.1 824 Intron XP_016873603.1
XM_017018115.1 824 Intron XP_016873604.1
XM_017018116.1 824 Intron XP_016873605.1
XM_017018117.1 824 Intron XP_016873606.1

View Full Product Details