Product Details

SNP ID
rs144707121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63508588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTCACGACGATTTTTGGAGGCCT[G/T]CATGCAGGCAAGATGGTCATGCTGC
Phenotype
MIM: 606096
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LGALS12 PubMed Links

Gene Details

Gene
LGALS12
Gene Name
galectin 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142535.1 512 Silent Mutation CTG,CTT L57L NP_001136007.1
NM_001142536.1 512 Silent Mutation CTG,CTT L57L NP_001136008.1
NM_001142537.1 512 UTR 5 NP_001136009.1
NM_001142538.1 512 UTR 5 NP_001136010.1
NM_033101.3 512 Silent Mutation CTG,CTT L57L NP_149092.2
XM_017018458.1 512 Silent Mutation CTG,CTT L57L XP_016873947.1

View Full Product Details