Product Details

SNP ID
rs144718261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:533776 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCACGGGGTTCACCTGTACTGGT[A/G]GATGTCCTCAAAAGACTTGGTGTTG
Phenotype
MIM: 190020
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HRAS PubMed Links

Gene Details

Gene
HRAS
Gene Name
HRas proto-oncogene, GTPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130442.2 485 Missense Mutation CAC,TAC H94Y NP_001123914.1
NM_001318054.1 485 UTR 5 NP_001304983.1
NM_005343.3 485 Missense Mutation CAC,TAC H94Y NP_005334.1
NM_176795.4 485 Missense Mutation CAC,TAC H94Y NP_789765.1
Gene
LOC101059906
Gene Name
collagen alpha-2(IV) chain-like
There are no transcripts associated with this gene.

Gene
LRRC56
Gene Name
leucine rich repeat containing 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198075.3 485 Intron NP_932341.1
XM_005252775.2 485 Intron XP_005252832.1
XM_005252776.2 485 Intron XP_005252833.1
XM_006718132.2 485 Intron XP_006718195.1
XM_006718133.2 485 Intron XP_006718196.1
XM_011519875.2 485 Intron XP_011518177.1
XM_011519876.1 485 Intron XP_011518178.1
XM_011519877.2 485 Intron XP_011518179.1
XM_017017167.1 485 Intron XP_016872656.1
XM_017017168.1 485 Intron XP_016872657.1

View Full Product Details