Product Details

SNP ID
rs144916716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:122089506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTAAATGCCAGCCACATGCAAAG[A/G]GGATGGAAAAGAAGAATGAGATTTG
Phenotype
MIM: 615965
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR100HG PubMed Links

Gene Details

Gene
MIR100HG
Gene Name
mir-100-let-7a-2 cluster host gene
There are no transcripts associated with this gene.

View Full Product Details