Product Details

SNP ID
rs145162136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62625969 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCCATAGTCATACCAATGGGCTA[C/T]AGCATAACAACAACGTTCCTACAGG
Phenotype
MIM: 606374 MIM: 104160
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B3GAT3 PubMed Links

Gene Details

Gene
B3GAT3
Gene Name
beta-1,3-glucuronyltransferase 3
There are no transcripts associated with this gene.

Gene
GANAB
Gene Name
glucosidase II alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278192.1 Intron NP_001265121.1
NM_001278193.1 Intron NP_001265122.1
NM_001278194.1 Intron NP_001265123.1
NM_198334.2 Intron NP_938148.1
NM_198335.3 Intron NP_938149.2
XM_017017412.1 Intron XP_016872901.1
XM_017017413.1 Intron XP_016872902.1
XM_017017414.1 Intron XP_016872903.1

View Full Product Details