Product Details
- SNP ID
-
rs145187500
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:73958360 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAGCACCACCCGTTGAAGTCAAAT[A/G]AGCCGTCTTCAGCAGAGATTTTCAG
- Phenotype
-
MIM: 610263
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DNAJB13
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141051090] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DNAJB13
- Gene Name
- DnaJ heat shock protein family (Hsp40) member B13
View Full Product Details