Product Details

SNP ID
rs145351881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6768026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCCTAGAGCGTGGAATGTGCC[A/G]GCAGTATGTATTTTCCCAGGACCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2AG2 PubMed Links

Gene Details

Gene
OR2AG2
Gene Name
olfactory receptor family 2 subfamily AG member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004490.1 Intron NP_001004490.1

View Full Product Details