Product Details

SNP ID
rs145947171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:70409954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTGGTGTGGAACAAGACCGAAT[A/G]GATAAGGTAAGTGGCCCGCGGCTGC
Phenotype
MIM: 164765
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTTN PubMed Links

Gene Details

Gene
CTTN
Gene Name
cortactin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184740.1 491 Missense Mutation ATA,ATG I95M NP_001171669.1
NM_005231.3 491 Missense Mutation ATA,ATG I95M NP_005222.2
NM_138565.2 491 Missense Mutation ATA,ATG I95M NP_612632.1
XM_006718447.2 491 Missense Mutation ATA,ATG I95M XP_006718510.1
XM_006718448.3 491 Missense Mutation ATA,ATG I95M XP_006718511.1
XM_017017312.1 491 Missense Mutation ATA,ATG I95M XP_016872801.1

View Full Product Details