Product Details

SNP ID
rs145994155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67399634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGTTGAAGCAGTCAGTGAAGGT[C/T]TTCCACAGTTTGATGTTGTAGCGTC
Phenotype
MIM: 176875 MIM: 603761 MIM: 610831
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100130987 PubMed Links

Gene Details

Gene
LOC100130987
Gene Name
uncharacterized LOC100130987
There are no transcripts associated with this gene.

Gene
PPP1CA
Gene Name
protein phosphatase 1 catalytic subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008709.1 606 Silent Mutation AAA,AAG K161K NP_001008709.1
NM_002708.3 606 Silent Mutation AAA,AAG K150K NP_002699.1
NM_206873.1 606 Silent Mutation AAA,AAG K106K NP_996756.1
Gene
RAD9A
Gene Name
RAD9 checkpoint clamp component A
There are no transcripts associated with this gene.

Gene
TBC1D10C
Gene Name
TBC1 domain family member 10C
There are no transcripts associated with this gene.

View Full Product Details