Product Details

SNP ID
rs146073659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73958404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCAGGCAAATAGCAGAGGCCTA[C/T]GACGTGCTGAGTGACCGTGAGTAGG
Phenotype
MIM: 610263
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJB13 PubMed Links

Gene Details

Gene
DNAJB13
Gene Name
DnaJ heat shock protein family (Hsp40) member B13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153614.3 1164 Silent Mutation TAC,TAT Y52Y NP_705842.2
XM_005273984.3 1164 Silent Mutation TAC,TAT Y52Y XP_005274041.1
XM_011545004.2 1164 Silent Mutation TAC,TAT Y86Y XP_011543306.1
XM_011545005.2 1164 Silent Mutation TAC,TAT Y86Y XP_011543307.1
XM_011545007.2 1164 UTR 5 XP_011543309.1
XM_011545009.2 1164 UTR 5 XP_011543311.1
XM_011545013.2 1164 Silent Mutation TAC,TAT Y86Y XP_011543315.1
XM_011545014.2 1164 Silent Mutation TAC,TAT Y86Y XP_011543316.1
XM_011545015.2 1164 Intron XP_011543317.1
XM_017017675.1 1164 UTR 5 XP_016873164.1
XM_017017676.1 1164 UTR 5 XP_016873165.1
XM_017017677.1 1164 UTR 5 XP_016873166.1
XM_017017678.1 1164 UTR 5 XP_016873167.1
XM_017017679.1 1164 Silent Mutation TAC,TAT Y86Y XP_016873168.1

View Full Product Details