Product Details

SNP ID
rs146283509
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66435418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTGAAGTAGCCCCTGGGATCTG[A/G]GTACTCCATGTAGTAACCCTGCCTG
Phenotype
MIM: 611826 MIM: 608554
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL11 PubMed Links

Gene Details

Gene
MRPL11
Gene Name
mitochondrial ribosomal protein L11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016050.4 1356 UTR 3 NP_057134.1
NM_170738.3 1356 UTR 3 NP_733934.1
NM_170739.3 1356 UTR 3 NP_733935.1
XM_017018135.1 1356 Intron XP_016873624.1
Gene
NPAS4
Gene Name
neuronal PAS domain protein 4
There are no transcripts associated with this gene.

View Full Product Details