Product Details

SNP ID
rs146463957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124543551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGGTTCCCCACCACGGTGACC[A/G]TGTAGAAACCCAGAAACAGGAAGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR8B12 PubMed Links

Gene Details

Gene
OR8B12
Gene Name
olfactory receptor family 8 subfamily B member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005195.1 104 Missense Mutation ACG,ATG T35M NP_001005195.1

View Full Product Details