Product Details

SNP ID
rs147081581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59503932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTTTTTCTCTCTCTGTGATGGC[A/G]TTTGACTGCTACATGGCCATCTCCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4D11 PubMed Links

Gene Details

Gene
OR4D11
Gene Name
olfactory receptor family 4 subfamily D member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004706.1 357 Silent Mutation GCA,GCG A119A NP_001004706.1

View Full Product Details