Product Details

SNP ID
rs147326331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62575522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGCCCCCGCTCTGCCAGGGCTG[C/T]GTGGCTGGGCTGCCCCTCTTCTCCA
Phenotype
MIM: 130593 MIM: 610641
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EEF1G PubMed Links

Gene Details

Gene
EEF1G
Gene Name
eukaryotic translation elongation factor 1 gamma
There are no transcripts associated with this gene.

Gene
MIR6747
Gene Name
microRNA 6747
There are no transcripts associated with this gene.

Gene
TUT1
Gene Name
terminal uridylyl transferase 1, U6 snRNA-specific
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022830.2 2349 Missense Mutation ACA,GCA T771A NP_073741.2

View Full Product Details