Product Details

SNP ID
rs147336691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8616265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAAGATGATCTCTCAGGTTGGGT[A/G]GAGAAGGAAATGAGCAATACCATCA
Phenotype
MIM: 612000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM66 PubMed Links

Gene Details

Gene
TRIM66
Gene Name
tripartite motif containing 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014818.1 6379 UTR 3 NP_055633.1
XM_006718397.2 6379 UTR 3 XP_006718460.1
XM_006718398.3 6379 UTR 3 XP_006718461.1
XM_011520504.1 6379 UTR 3 XP_011518806.1
XM_011520507.2 6379 UTR 3 XP_011518809.1
XM_011520508.1 6379 UTR 3 XP_011518810.1
XM_011520509.1 6379 UTR 3 XP_011518811.1
XM_011520510.1 6379 UTR 3 XP_011518812.1
XM_011520511.1 6379 UTR 3 XP_011518813.1
XM_011520512.1 6379 UTR 3 XP_011518814.1
XM_011520513.1 6379 UTR 3 XP_011518815.1
XM_011520514.2 6379 UTR 3 XP_011518816.1
XM_011520515.1 6379 UTR 3 XP_011518817.1
XM_011520516.1 6379 UTR 3 XP_011518818.1
XM_011520517.2 6379 UTR 3 XP_011518819.1
XM_011520518.1 6379 UTR 3 XP_011518820.1
XM_011520519.1 6379 UTR 3 XP_011518821.1
XM_011520522.2 6379 UTR 3 XP_011518824.1
XM_011520523.2 6379 UTR 3 XP_011518825.1
XM_011520524.1 6379 Intron XP_011518826.1
XM_011520525.1 6379 Intron XP_011518827.1
XM_011520526.2 6379 UTR 3 XP_011518828.1
XM_011520527.1 6379 UTR 3 XP_011518829.1
XM_017018629.1 6379 UTR 3 XP_016874118.1
XM_017018630.1 6379 UTR 3 XP_016874119.1
XM_017018631.1 6379 UTR 3 XP_016874120.1

View Full Product Details